Please login to your online account to display your discounted pricing

Mouse anti-pan Actin, Clone: SPM160, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439902

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Rat
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Isotype IgG1, κ
Format Purified
Clone SPM160
Purification Method Purified
Immunogen SDS extract of human myocardium.
Host Species Mouse
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Antigen pan Actin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Concentration 1mg/mL
Test Specificity This antibody recognizes actin of skeletal, cardiac, and smooth muscle cells. It is not reactive with other mesenchymal cells except for myoepithelium.
Regulatory Status RUO
Gene ID (Entrez) 58