Please login to your online account to display your discounted pricing

Mouse anti-pan Actin, Clone: HHF35, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen pan Actin
Clone HHF35
Concentration 0.2mg/mL
Immunogen SDS extract of human myocardium.
Gene Accession No. P62736
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23423001

 
novus biologicals canada
NBP23423001ML
0.1mL Each for $139.00

NBP23423005

 
novus biologicals canada
NBP23423005ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen pan Actin
Clone HHF35
Concentration 0.2mg/mL
Immunogen SDS extract of human myocardium.
Gene Accession No. P62736
Gene ID (Entrez) 58
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting