Please login to your online account to display your discounted pricing

Mouse anti-pan Actin, Clone: HHF35, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423005

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen pan Actin
Quantity 0.5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen SDS extract of human myocardium.
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 58
Reconstitution Protein A purified
Clone HHF35
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Rat
Gene Accession No. P62736
Purification Method Purified
Concentration 0.2mg/mL