Please login to your online account to display your discounted pricing

Mouse anti-pan Actin, Clone: HHF35, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463401

  • / Each
 

Description & Specifications

Specifications

Antigen pan Actin
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone HHF35
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P62736
Host Species Murine
Immunogen SDS extract of human myocardium.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 58
Reconstitution Protein A purified
Target Species Human
Target Species Rabbit
Target Species Rat