Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: SPM590, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438705

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen p53
Monoclonal or Polyclonal Monoclonal
Format Purified
Quantity 0.5mL
Immunogen Recombinant human wild type p53 protein expressed in E. coli.
Primary or Secondary Primary
Gene ID (Entrez) 7157
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL
Host Species Mouse
Conjugate Unlabeled
Regulatory Status RUO
Test Specificity Recognizes a 53kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53. Its epitope maps within the N-terminus (aa 37-45) of p53.
Gene Accession No. P04637
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Isotype IgG2b, κ
Clone SPM590