Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: SPM590, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438701

  • / Each
 

Description & Specifications

Specifications

Antigen p53
Format Purified
Immunogen Recombinant human wild type p53 protein expressed in E. coli.
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Formulation PBS with 0.05% BSA.
Gene Accession No. P04637
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 7157
Test Specificity Recognizes a 53kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53. Its epitope maps within the N-terminus (aa 37-45) of p53.
Reconstitution Protein A purified
Clone SPM590
Concentration 0.2mg/mL
Conjugate Unlabeled
Primary or Secondary Primary