Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: SPM590, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23443402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23443402

  • / Each
 

Description & Specifications

Specifications

Antigen p53
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P04637
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.2mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7157
Test Specificity Recognizes a 53kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53. Its epitope maps within the N-terminus (aa 37-45) of p53.
Reconstitution Protein A purified
Clone SPM590
Concentration 1mg/mL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Immunogen Recombinant human wild type p53 protein expressed in E. coli.
Isotype IgG2b, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled