Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: SPM590, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23443401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23443401

  • / Each
 

Description & Specifications

Specifications

Antigen p53
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM590
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P04637
Host Species Murine
Immunogen Recombinant human wild type p53 protein expressed in E. coli.
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7157
Test Specificity Recognizes a 53kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53. Its epitope maps within the N-terminus (aa 37-45) of p53.
Reconstitution Protein A purified
Target Species Bovine
Target Species Human
Target Species Primate