Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: SPM589, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen p53
Clone SPM589
Concentration 0.2mg/mL
Immunogen Recombinant human wild-type p53 protein
Gene Accession No. P04637
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23289101

 
novus biologicals canada
NBP23289101ML
0.1mL Each for $139.00

NBP23289110

 
novus biologicals canada
NBP23289110ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen p53
Clone SPM589
Concentration 0.2mg/mL
Immunogen Recombinant human wild-type p53 protein
Gene Accession No. P04637
Gene ID (Entrez) 7157
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting