Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: SPM589, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23289101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23289101

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Antigen p53
Format Purified
Host Species Mouse
Immunogen Recombinant human wild-type p53 protein
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 7157
Test Specificity Does not react to Mouse and Rat.
Reconstitution Protein A purified
Clone SPM589
Concentration 0.2mg/mL
Conjugate Unlabeled
Gene Accession No. P04637
Isotype IgG2a