Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: SPM589, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23481701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23481701

  • / Each
 

Description & Specifications

Specifications

Antigen p53
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM589
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P04637
Host Species Murine
Immunogen Recombinant human wild-type p53 protein
Isotype IgG2a
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7157
Test Specificity Recognizes a 53kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53 under denaturing and non-denaturing conditions. Its epitope maps within the N-terminus (aa 20-25) of p53 oncoprotein. Does not react with Mouse or Rat.
Reconstitution Protein A purified
Target Species Human