Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: PAb122, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen p53
Clone PAb122
Concentration 1mg/mL
Immunogen SV40-transformed mouse B4 cells
Gene Accession No. P04637
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23470401

 
novus biologicals canada
NBP23470401MG
0.1mg Each for $384.00

NBP23470402

 
novus biologicals canada
NBP23470402MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen p53
Clone PAb122
Concentration 1mg/mL
Immunogen SV40-transformed mouse B4 cells
Gene Accession No. P04637
Gene ID (Entrez) 7157
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting