Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: PAb122, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470402

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Regulatory Status RUO
Purification Method Purified
Concentration 1mg/mL
Quantity 0.2mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG2b, κ
Antigen p53
Gene ID (Entrez) 7157
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P04637
Clone PAb122
Host Species Mouse
Immunogen SV40-transformed mouse B4 cells