Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: PAb122, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470401

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen SV40-transformed mouse B4 cells
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P04637
Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Gene ID (Entrez) 7157
Antigen p53
Purification Method Purified
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Conjugate Unlabeled
Clone PAb122
Isotype IgG2b, κ
Reconstitution Protein A purified