Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: DO-7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23430805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23430805

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Immunogen Recombinant human wild type p53 protein expressed in E. coli.
Regulatory Status RUO
Format Purified
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Antigen p53
Gene Accession No. P04637
Isotype IgG2b, κ
Purification Method Purified
Gene ID (Entrez) 7157
Concentration 0.2mg/mL
Clone DO-7