Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: DO-7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23430801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23430801

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7157
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Antigen p53
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P04637
Immunogen Recombinant human wild type p53 protein expressed in E. coli.
Isotype IgG2b, κ
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone DO-7
Concentration 0.2mg/mL