missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: DO-7, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen p53
Clone DO-7
Concentration 1mg/mL
Immunogen Recombinant human wild type p53 protein expressed in E. coli.
Gene Accession No. P04637
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23460501 Novus Biologicals Canada
NBP23460501MG
0.1mg Each for $384.00
NBP23460502 Novus Biologicals Canada
NBP23460502MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

p53
1mg/mL
P04637
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
DO-7
Recombinant human wild type p53 protein expressed in E. coli.
7157
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot