Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: DO-7, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23460501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23460501

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Isotype IgG2b, κ
Gene ID (Entrez) 7157
Reconstitution Protein A purified
Gene Accession No. P04637
Quantity 0.1mg
Regulatory Status RUO
Antigen p53
Immunogen Recombinant human wild type p53 protein expressed in E. coli.
Purification Method Purified
Clone DO-7
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Concentration 1mg/mL