Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: BP53-12 + DO-7, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23430905ML

 View all options for this product


For Research Use Only

Catalog No. NBP23430905


Description & Specifications


Antigen p53
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone BP53-12 + DO-7
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P04637
Host Species Mouse
Immunogen Recombinant human wild-type p53 protein (BP53-12) and Recombinant human wild type p53 protein expressed in E. coli (DO-7)
Isotype IgG2a
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7157
Reconstitution Protein A purified