Please login to your online account to display your discounted pricing

Mouse anti-p53, Clone: BP53-12 + DO-7, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23456301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23456301

  • / Each
 

Description & Specifications

Specifications

Antigen p53
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone BP53-12 + DO-7
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P04637
Host Species Murine
Immunogen Recombinant human wild-type p53 protein (BP53-12) and Recombinant human wild type p53 protein expressed in E. coli (DO-7)
Isotype IgG2a
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7157
Reconstitution Protein A purified
Target Species Bovine
Target Species Human
Target Species Primate