Please login to your online account to display your discounted pricing

Mouse anti-p27/Kip1, Clone: SX53G8, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen p27/Kip1
Clone SX53G8
Concentration 1mg/mL
Immunogen Purified GST-p27 fusion protein of human origin
Gene Accession No. P46527
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23471801

 
novus biologicals canada
NBP23471801MG
0.1mg Each for $384.00

NBP23471802

 
novus biologicals canada
NBP23471802MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen p27/Kip1
Clone SX53G8
Concentration 1mg/mL
Immunogen Purified GST-p27 fusion protein of human origin
Gene Accession No. P46527
Gene ID (Entrez) 1027
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting