Please login to your online account to display your discounted pricing

Mouse anti-p27/Kip1, Clone: SX53G8, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471801

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 1027
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Quantity 0.1mg
Reconstitution Protein A purified
Gene Accession No. P46527
Clone SX53G8
Purification Method Purified
Antigen p27/Kip1
Host Species Mouse
Format Purified
Monoclonal or Polyclonal Monoclonal
Immunogen Purified GST-p27 fusion protein of human origin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Isotype IgG1, κ
Regulatory Status RUO