Please login to your online account to display your discounted pricing

Mouse anti-p27/Kip1, Clone: SX53G8, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471802

  • / Each
 

Description & Specifications

Specifications

Antigen p27/Kip1
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Gene ID (Entrez) 1027
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P46527
Immunogen Purified GST-p27 fusion protein of human origin
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone SX53G8
Concentration 1mg/mL