Please login to your online account to display your discounted pricing

Mouse anti-p27/Kip1, Clone: SPM348, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23289010ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23289010

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Antigen p27/Kip1
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P46527
Host Species Mouse
Immunogen Purified GST-p27 fusion protein of human origin
Isotype IgG1, κ
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1027
Purification Method Purified
Regulatory Status RUO
Reconstitution Protein A purified
Clone SPM348
Concentration 0.2mg/mL
Conjugate Unlabeled