Please login to your online account to display your discounted pricing

Mouse anti-p27/Kip1, Clone: SPM348, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476602

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1027
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen p27/Kip1
Isotype IgG1, κ
Primary or Secondary Primary
Conjugate Unlabeled
Format Purified
Clone SPM348
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Purified GST-p27 fusion protein of human origin
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.2mg
Regulatory Status RUO
Test Specificity This antibody recognizes a 27kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. It is highly specific and shows no cross-reaction with other related mitotic inhibitors.
Concentration 1mg/mL
Reconstitution Protein A purified
Gene Accession No. P46527