Please login to your online account to display your discounted pricing

Mouse anti-p27/Kip1, Clone: SPM348, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476601

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Antigen p27/Kip1
Gene Accession No. P46527
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified GST-p27 fusion protein of human origin
Isotype IgG1, κ
Quantity 0.1mg
Primary or Secondary Primary
Test Specificity This antibody recognizes a 27kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. It is highly specific and shows no cross-reaction with other related mitotic inhibitors.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Purification Method Purified
Regulatory Status RUO
Gene ID (Entrez) 1027
Reconstitution Protein A purified
Clone SPM348
Concentration 1mg/mL
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal