Please login to your online account to display your discounted pricing

Mouse anti-P-Selectin/CD62P, Clone: Psel.KO2.3, PE, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen P-Selectin/CD62P
Clone Psel.KO2.3
Immunogen The immunogen for this antibody was CD62P.
Gene ID (Entrez) 6403
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14385201

 
novus biologicals canada
NBP14385201MG
0.1mg Each for $252.00

NBP14385203

 
novus biologicals canada
NBP1438520025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen P-Selectin/CD62P
Clone Psel.KO2.3
Immunogen The immunogen for this antibody was CD62P.
Gene ID (Entrez) 6403
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry