Please login to your online account to display your discounted pricing

Mouse anti-P-Selectin/CD62P, Clone: AK-4, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen P-Selectin/CD62P
Clone AK-4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD62P.
Gene ID (Entrez) 6403
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14339201

 
novus biologicals canada
NBP14339201MG
0.1mg Each for $285.00

NBP14339203

 
novus biologicals canada
NBP1433920025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen P-Selectin/CD62P
Clone AK-4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD62P.
Gene ID (Entrez) 6403
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)