Please login to your online account to display your discounted pricing

Mouse anti-OX40 Ligand/TNFSF4, Clone: ATM2, Novus Biologicals™

$274.00 - $307.00

Specifications

Antigen OX40 Ligand/TNFSF4
Clone ATM2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD134L.
Gene ID (Entrez) 7292
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14335301

 
novus biologicals canada
NBP14335301MG
0.1mg Each for $307.00

NBP14335305

 
novus biologicals canada
NBP143353005MG
0.05mg Each for $274.00
Description & Specifications

Specifications

Antigen OX40 Ligand/TNFSF4
Clone ATM2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD134L.
Gene ID (Entrez) 7292
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Blocking
Applications Flow Cytometry
Applications Neutralization