Please login to your online account to display your discounted pricing

Mouse anti-OX40 Ligand/TNFSF4, Clone: ATM2, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14335301MG

 View all options for this product


For Research Use Only

Catalog No. NBP14335301


Description & Specifications


Antigen OX40 Ligand/TNFSF4
Applications Blocking
Applications Flow Cytometry
Applications Neutralization
Clone ATM2
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Rat
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Immunogen The immunogen for this antibody was CD134L.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7292