Please login to your online account to display your discounted pricing

Mouse anti-OX40 Ligand/TNFSF4, Clone: ATM2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143353005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14335305

  • / Each
 

Description & Specifications

Specifications

Antigen OX40 Ligand/TNFSF4
Applications Blocking
Applications Flow Cytometry
Applications Neutralization
Primary or Secondary Primary
Cross Reactivity Rat
Format Affinity Purified
Host Species Mouse
Immunogen The immunogen for this antibody was CD134L.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7292
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation Phosphate buffer pH 7.2
Clone ATM2