Please login to your online account to display your discounted pricing

Mouse anti-OSMR, Clone: AN-V2, PE, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen OSMR
Clone AN-V2
Immunogen The immunogen for this antibody is OSMR.
Gene ID (Entrez) 9180
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20033625

 
novus biologicals canada
NBP20033625TESTS
25 Tests Each for $175.00

NBP200336

 
novus biologicals canada
NBP200336100TESTS
100 Tests Each for $362.00
Description & Specifications

Specifications

Antigen OSMR
Clone AN-V2
Immunogen The immunogen for this antibody is OSMR.
Gene ID (Entrez) 9180
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry