Please login to your online account to display your discounted pricing

Mouse anti-Ornithine Decarboxylase, Clone: SPM565, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23288810ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23288810

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SPM565
Conjugate Unlabeled
Cross Reactivity Human
Antigen Ornithine Decarboxylase
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Recombinant human ODC-1 protein
Isotype IgG1, κ
Purification Method Purified
Gene ID (Entrez) 4953
Test Specificity Recognizes a 53kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.
Reconstitution Protein A purified
Concentration 0.2mg/mL
Gene Accession No. P11926
Regulatory Status RUO