Please login to your online account to display your discounted pricing

Mouse anti-Ornithine Decarboxylase, Clone: SPM565, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Ornithine Decarboxylase
Clone SPM565
Concentration 1mg/mL
Immunogen Recombinant human ODC-1 protein
Gene Accession No. P11926
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23480001

 
novus biologicals canada
NBP23480001MG
0.1mg Each for $384.00

NBP23480002

 
novus biologicals canada
NBP23480002MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Ornithine Decarboxylase
Clone SPM565
Concentration 1mg/mL
Immunogen Recombinant human ODC-1 protein
Gene Accession No. P11926
Gene ID (Entrez) 4953
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot