Please login to your online account to display your discounted pricing

Mouse anti-OATP1B3/SLCO1B3/OATP8, Clone: 1F6.1B1, Novus Biologicals™

$109.00 - $259.02

Specifications

Antigen OATP1B3/SLCO1B3/OATP8
Clone 1F6.1B1
Concentration 1.0mg/mL
Immunogen Two synthetic peptides made to the human SLCO1B3 protein sequence (between residues 648-700). [Swiss-Prot Q9NPD5]
Gene Accession No. Q9NPD5
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP236493SS

 
novus biologicals canada
NBP236493SS
0.025mg Each for $109.00

NBP236493

 
novus biologicals canada
NBP236493
0.1mg Each for $259.02
Description & Specifications

Specifications

Antigen OATP1B3/SLCO1B3/OATP8
Clone 1F6.1B1
Concentration 1.0mg/mL
Immunogen Two synthetic peptides made to the human SLCO1B3 protein sequence (between residues 648-700). [Swiss-Prot Q9NPD5]
Gene Accession No. Q9NPD5
Gene ID (Entrez) 28234
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)