Please login to your online account to display your discounted pricing

Mouse anti-OATP1B3/SLCO1B3/OATP8, Clone: 1E6.2F9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP236492SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP236492SS

  • / Each
 

Description & Specifications

Specifications

Antigen OATP1B3/SLCO1B3/OATP8
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone 1E6.2F9
Concentration 1.0mg/mL
Conjugate Unlabeled
Format Purified
Formulation PBS. Protein G purified.
Gene Accession No. Q9NPD5
Host Species Murine
Immunogen Two synthetic peptides made to the human SLCO1B3 protein sequence (between residues 648-700). [Swiss-Prot Q9NPD5]
Purification Method Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 28234
Target Species Human