Please login to your online account to display your discounted pricing

Mouse anti-Nucleoli Marker, Clone: NM95, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Nucleoli Marker
Clone NM95
Concentration 1mg/mL
Immunogen Nuclei of myeloid leukemia biopsy cells
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23469501

 
novus biologicals canada
NBP23469501MG
0.1mg Each for $384.00

NBP23469502

 
novus biologicals canada
NBP23469502MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Nucleoli Marker
Clone NM95
Concentration 1mg/mL
Immunogen Nuclei of myeloid leukemia biopsy cells
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence