Please login to your online account to display your discounted pricing

Mouse anti-Nucleoli Marker, Clone: NM95, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469501

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Primary or Secondary Primary
Regulatory Status RUO
Cross Reactivity Human
Concentration 1mg/mL
Immunogen Nuclei of myeloid leukemia biopsy cells
Isotype IgG1, κ
Quantity 0.1mg
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone NM95
Purification Method Purified
Host Species Mouse
Conjugate Unlabeled
Antigen Nucleoli Marker
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.