Please login to your online account to display your discounted pricing

Mouse anti-Nuclear Membrane Marker, Clone: NM97, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469601

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Regulatory Status RUO
Format Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Purification Method Purified
Concentration 1mg/mL
Primary or Secondary Primary
Clone NM97
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Immunogen Nuclei of myeloid leukemia biopsy cells
Antigen Nuclear Membrane Marker