Please login to your online account to display your discounted pricing

Mouse anti-Nuclear Membrane Marker, Clone: NM97, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469602

  • / Each
 

Description & Specifications

Specifications

Antigen Nuclear Membrane Marker
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Quantity 0.2mg
Format Purified
Immunogen Nuclei of myeloid leukemia biopsy cells
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone NM97
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human