Please login to your online account to display your discounted pricing

Mouse anti-Nuclear Antigen, Clone: 235-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434205

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Antigen Nuclear Antigen
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Nuclei of human myeloid leukemia biopsy cells
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Does not react to Chicken, Mouse or Rat.
Reconstitution Protein A purified
Clone 235-1
Conjugate Unlabeled
Cross Reactivity Human
Quantity 0.5mL
Concentration 0.2mg/mL