Please login to your online account to display your discounted pricing

Mouse anti-Nuclear Antigen, Clone: 235-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434201

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity Does not react to Chicken, Mouse or Rat.
Cross Reactivity Human
Antigen Nuclear Antigen
Format Purified
Formulation PBS with 0.05% BSA.
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Immunogen Nuclei of human myeloid leukemia biopsy cells
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone 235-1
Concentration 0.2mg/mL
Conjugate Unlabeled