missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Nuclear Antigen, Clone: 235-1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Nuclear Antigen
Clone 235-1
Concentration 1mg/mL
Immunogen Nuclei of human myeloid leukemia biopsy cells
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23452501 Novus Biologicals Canada
NBP23452501MG
0.1mg Each for $384.00
NBP23452502 Novus Biologicals Canada
NBP23452502MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Nuclear Antigen
1mg/mL
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
235-1
Nuclei of human myeloid leukemia biopsy cells
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Immunocytochemistry,Immunofluorescence,Immunoprecipitation