Please login to your online account to display your discounted pricing

Mouse anti-Nuclear Antigen, Clone: 235-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23452501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23452501

  • / Each
 

Description & Specifications

Specifications

Format Purified
Isotype IgG1, κ
Reconstitution Protein A purified
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Antigen Nuclear Antigen
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Does not react to Chicken, Mouse or Rat.
Conjugate Unlabeled
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Immunogen Nuclei of human myeloid leukemia biopsy cells
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone 235-1
Concentration 1mg/mL
Purification Method Purified
Quantity 0.1mg