Please login to your online account to display your discounted pricing

Mouse anti-Nuclear Antigen, Clone: 235-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23452502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23452502

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Antigen Nuclear Antigen
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Isotype IgG1, κ
Primary or Secondary Primary
Test Specificity Does not react to Chicken, Mouse or Rat.
Cross Reactivity Human
Immunogen Nuclei of human myeloid leukemia biopsy cells
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone 235-1
Concentration 1mg/mL
Conjugate Unlabeled