Please login to your online account to display your discounted pricing

Mouse anti-NKX2.2, Clone: SPM564, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen NKX2.2
Clone SPM564
Concentration 0.2mg/mL
Immunogen Human NKX2.2 recombinant protein
Gene Accession No. O95096
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23288401

 
novus biologicals canada
NBP23288401ML
0.1mL Each for $139.00

NBP23288410

 
novus biologicals canada
NBP23288410ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen NKX2.2
Clone SPM564
Concentration 0.2mg/mL
Immunogen Human NKX2.2 recombinant protein
Gene Accession No. O95096
Gene ID (Entrez) 4821
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting