Please login to your online account to display your discounted pricing

Mouse anti-NKX2.2, Clone: SPM564, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23288410ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23288410

  • / Each
 

Description & Specifications

Specifications

Antigen NKX2.2
Immunogen Human NKX2.2 recombinant protein
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation PBS with 0.05% BSA.
Clone SPM564
Host Species Mouse
Format Purified
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Quantity 1.0mL
Isotype IgG2b, κ
Gene Accession No. O95096
Concentration 0.2mg/mL
Gene ID (Entrez) 4821
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Purification Method Purified