Please login to your online account to display your discounted pricing

Mouse anti-NKX2.2, Clone: SPM564, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23288401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23288401

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen NKX2.2
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. O95096
Immunogen Human NKX2.2 recombinant protein
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4821
Reconstitution Protein A purified
Clone SPM564
Concentration 0.2mg/mL
Conjugate Unlabeled
Host Species Mouse