missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-NKX2.2, Clone: SPM564, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen NKX2.2
Clone SPM564
Concentration 1mg/mL
Immunogen Human NKX2.2 recombinant protein
Gene Accession No. O95096
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23479901 Novus Biologicals Canada
NBP23479901MG
0.1mg Each for $384.00
NBP23479902 Novus Biologicals Canada
NBP23479902MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

NKX2.2
1mg/mL
O95096
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM564
Human NKX2.2 recombinant protein
4821
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot