Please login to your online account to display your discounted pricing

Mouse anti-NKX2.2, Clone: SPM564, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479902

  • / Each
 

Description & Specifications

Specifications

Quantity 0.2mg
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Mouse
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 4821
Regulatory Status RUO
Immunogen Human NKX2.2 recombinant protein
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Purification Method Purified
Clone SPM564
Gene Accession No. O95096
Concentration 1mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Isotype IgG2b, κ
Antigen NKX2.2